Learn More
Hach Company APA6000 Copper Standard 2 (High Range), 10.0 mg/L, 1 L
Supplier:  Hach Company 2756453
Encompass
High-range copper standard solution at 10.0 mg/L supplied in a 1 L bottle for calibration and quality control in water and wastewater analysis.
  • Ready-to-use aqueous standard for instrument calibration and method verification.
  • Accurate 10.0 mg/L concentration for high-range copper measurements.
  • Suitable for calibration of spectrophotometers and online analyzers.
  • Supplied in a single 1 L bottle for laboratory use.
  • Safety Data Sheet available; handle according to laboratory safety procedures.
Catalog No. NC2309992