Learn More
Cayman Chemical HR2RbB2 ChImRIc MoncLNa 200ug
SDP
Supplier:  Cayman Chemical 37168200 UG
Encompass_Preferred
Immunogen: A431 cells overexpressing EGFR • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) Human • Cross Reactivity: (+) HER2 • Applications: ELISA, FC, IHC
Catalog No. 50-291-4794