Learn More
Cayman Chemical HR2RBB2 EXrAClular Doma 100ug
SDP
Supplier:  Cayman Chemical 32007100 UG
Encompass_Preferred
Source: Active recombinant C-terminal human IgG1 Fc-tagged HER/ERBB2 expressed in HEK293 cells • Amino acids: 23-652 • MW: 96.1 kDa
Catalog No. 50-294-8934