Learn More
Vector Laboratories HRP Goat Anti-Rabbit IgG Antibody (Peroxidase) 1 mg
SDP
Supplier:  Vector Laboratories PI10001
Encompass
Vector Laboratories secondary antibodies are prepared using proprietary immunization schedules that produce high affinity antibodies. The antibodies are then purified by affinity chromatography, and cross-reactivities that are likely to interfere with specific labeling are removed by solid phase adsorption techniques. This HRP Goat Anti-Rabbit IgG Antibody is cross-linked with high specific activity horseradish peroxidase. The antibodies are subjected to rigorous quality control assays and can be used for tissue and cell staining, ELISAs, and blots.
Catalog No. NC9555586