Learn More
Seracare Life Sciences Inc HRP CHEMI NA BLOTTING KIT
Supplier:  Seracare Life Sciences Inc 59100027
Encompass
The Detector HRP Chemiluminsescent Kit is designed for non-radioactive detection of membrane-bound DNA using biotinylated DNA probes, peroxidase linked streptavidin, and chemiluminescent substrate. The rapid detection procedure produces results in about 1.5 hours. The kit contains all reagents necessary to perform hybridization and detection of membrane bound DNA, except for the biotinylated probe. Applications include Southern blotting, bacterial colony hybridizations, plaque hybridizations, and dot blotting. Key benefits include detection of 0.3 pg DNA after just 15 minutes with minimal background; hence optimal signal to noise ratios and blots can be stripped and reprobed.
Catalog No. 50-675-05