Learn More
ABclonal Technology HRP conjugated Anti- IgG2b Fc
SDP
Supplier:  ABclonal Technology AS123100UL
Encompass_Preferred
Enables antigen binding activity Acts upstream of or within phagocytosis positive regulation of immune response and positive regulation of phagocytosis Located in extracellular space Part of immunoglobulin complex circulating Orthologous to several human genes including IGHG1 (immunoglobulin heavy constant gamma 1 (G1m marker)) IGHG2 (immunoglobulin heavy constant gamma 2 (G2m marker)) and IGHG3 (immunoglobulin heavy constant gamma 3 (G3m marker))
Catalog No. 50-000-13636