Learn More
ABclonal Technology HRP-conjugated B-Tubulin Mouse
SDP
Supplier:  ABclonal Technology AC03050UL
Encompass_Preferred
This gene encodes a beta tubulin protein This protein forms a dimer with alpha tubulin and acts as a structural component of microtubules Mutations in this gene cause cortical dysplasia complex with other brain malformations 6 Alternative splicing results in multiple splice variants There are multiple pseudogenes for this gene on chromosomes 1 6 7 8 9 and 13
Catalog No. 50-000-15012