Learn More
Medchemexpress LLC HRP-conjugated Goat Anti-Rat IgG H&L | 100 UL

Supplier: Medchemexpress LLC HYP83751100UL
HRP-conjugated Goat Anti-Rat IgG H&L is an HRP-conjugated antibody originating from a Goat, specifically targeting the Rat IgG antibody. It is intended for research use only.
- Goat host
- Polyclonal clonality
- Reacts with rat species
- Immunogen: Rat IgG(H+L)
- Affinity purified
- HRP conjugated
- IgG isotype
- Liquid appearance
- Formulated in 1*TBS (pH7.4), 0.5%BSA, 40% Glycerol
- Applications include Western Blot (WB) and Enzyme Linked Immunosorbent Assay (ELISA)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.