Learn More
Abcam Mouse and Rabbit Specific HRP/DAB Detection IHC kit, 15ML
Supplier:  Abcam AB6426415ML
Encompass_Preferred
HRP/DAB detection IHC kit, mouse & rabbit specific. Is a complete kit to detect a primary antibody raised in either mouse or rabbit, using HRP-labeled-streptavidin and a biotinylated anti-rabbit/anti-mouse secondary antibody. - Cited in over 220 publications - HRP produces a brown colored substance at the site of primary antibody binding by reacting with DAB
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0694127