Learn More
Apexbio Technology LLC HRP Goat Anti-Rabbit IgG (H L) Antibody 1ml
SDP
Supplier:  Apexbio Technology LLC K12231
Encompass_Preferred
Anti-rabbit secondary antibodies are affinity-purified antibodies with specificity for rabbit immunoglobulins suitable for detection sorting or purification of their specific targets Secondary antibodies provide additional functionalities allowing users to utilize various detection systems (e g HRP AP fluorescence) They can also offer enhanced sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody Typically secondary antibodies are generated by immunizing a host animal with an immunoglobulin pool from the target species These antibodies can be further purified and modified (such as by immunoaffinity chromatography antibody fragmentation conjugation labeling etc ) to produce highly specific reagents
Catalog No. 50-310-7443