Learn More
Diagnostic Biosystems HRP PERM RED CHROMOGEN - 30MLS PermaRed™/HRP
Supplier:  Diagnostic Biosystems K075
Encompass_Preferred
PermaRed™/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase. PermaRed™/HRP can be permanently mounted to produce a strong red brick color that can be easily distinguished from other stains.
Catalog No. NC1207405