Learn More
Biolegend HRP Streptavidin Reactivity: Human, Mouse, Rat, All Species; Apps: ELISA, ELISPOT, IHC, WB; Size: 1 mL
Supplier:  Biolegend 405210
Encompass_Preferred
HRP Streptavidin Reactivity: Human, Mouse, Rat, All Species; Apps: ELISA, ELISPOT, IHC, WB; Size: 1 mL
Catalog No. 50-169-081