Learn More
ATCC Hs 936.T(C1) cells (ATCC CRL-7687) - human melanoma skin epithelial adherent cell line
Supplier:  ATCC CRL7687
Encompass
Hs 936.T(C1) (CRL-7687) is a human skin melanoma-derived epithelial cell line provided for in vitro research applications. The line grows as adherent monolayers and is maintained under standard mammalian culture conditions in Dulbecco's Modified Eagle's Medium with serum.
  • Human skin melanoma-derived epithelial cell line.
  • Adherent growth suitable for monolayer culture.
  • Maintained in DMEM with 10% fetal bovine serum.
  • Culture at 37°C with 5% CO2; medium renewal every 2-3 days.
  • Subculture ratio 1:2 to 1:4 for routine passaging.
  • Suitable for in vitro melanoma biology and drug response assays.
  • Cryopreserved for storage in vapor phase liquid nitrogen.
Catalog No. NC3915223