Learn More
Enzo Life Sciences AMPINEXT™ High-Sensitivity Bisulfite-Seq Kit (Illumina) (12Reactions)
SDP
Supplier:  Enzo Life Sciences ENZGEN5080012
Encompass_Preferred
The AMPINEXT™ High-Sensitivity Bisulfite-Seq Kit (Illumina) is designed to bisulfite-convert DNA and prepare an Illumina-based library for bisulfite sequencing, all in one kit. Intended applications include whole genome (WGBS), oxidative (oxBS-seq), reduced representation (RRBS), and other bisulfite-next generation sequencing. The optimized protocol and components of the kit allow subnanogram amounts of DNA to be  bisulfite converted and fragmented simultaneously followed by quick non-barcoded (singleplexed) and barcoded (multiplexed) library construction in less than 7 hours. Assay Time: <7 hours.
Catalog No. 50-201-3794