Learn More
Trajan Scientific Headspace Magnetic Crimp Cap 20mm, 8mm hole, PTFE/Clear Silicone, 3mm, 100 Pack
Supplier:  Trajan Scientific XAR20039608
Encompass_Preferred
Cap, Crimp, Headspace, Magnetic With 8mm hole, 0.005 in. PTFE/Clear Silicone - 3.2mm, 100 Pack
Catalog No. 50-635-125