Learn More
Medchemexpress LLC HS-PEG5-CH2CH2NH2 hydrochloride | 1446256-16-1 | 98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY130872A
Encompass_Preferred
HS-PEG5-CH2CH2NH2 hydrochloride is a PROTAC linker belonging to the PEG class, used for synthesizing PROTAC molecules. It is stable under recommended storage conditions and classified as not a hazardous substance or mixture.
  • Used for synthesizing PROTAC molecules
  • PROTAC linker belonging to the PEG class
  • Stable under recommended storage conditions
  • Not classified as a hazardous substance or mixture
Catalog No. 50-003-41420