Learn More
Medchemexpress LLC Hs-peg6-ch2ch2-boc | 1818294-40-4 | MFCD26127815 | 95.0% | 426.57 g/mol | C19H38O8S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1308815MG
Encompass_Preferred
HS-PEG6-CH2CH2-Boc is a PEG-based linker bearing a thiol terminus and a Boc-protected terminus, designed for use in the synthesis of PROTACs and other bifunctional molecules. It provides a flexible, hydrophilic spacer useful for connecting ligands while maintaining solubility and synthetic handle orthogonality.
  • Suitable for use in PROTAC and bifunctional molecule synthesis.
  • Thiol terminus enables selective conjugation to thiol-reactive partners.
  • PEG6 spacer provides flexibility and improved aqueous solubility.
  • Boc-protected terminus allows orthogonal protection and selective deprotection.
  • Molecular formula C19H38O8S; molecular weight 426.57 g/mol.
Catalog No. 50-002-63965