Learn More
Medchemexpress LLC HS024 TFA | 99.80% | 1380.52 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP1215A5MG
Encompass_Preferred
HS024 is a selective antagonist for the MC4 receptor, exhibiting binding affinities (Ki) of 0.29 nM for MC4, 3.29 nM for MC5, 5.45 nM for MC3, and 18.6 nM for MC1. This compound has been shown to increase food intake, suggesting its potential role in appetite regulation studies.
  • Selective MC4 receptor antagonist.
  • Increases food intake.
  • Suitable for in vitro and in vivo research.
  • High purity.
Catalog No. 50-004-23337