Learn More
Medchemexpress LLC hsa-miR-296-3p agomi 20nmol | 20nmol
SDP
Supplier:  Medchemexpress LLC HYR00505A20NMOL
Encompass_Preferred
hsa-miR-296-3p agomirs are chemically-modified double-strand miRNA mimics with modified mature miRNA strand 2 phosphorothioates at the 5 end 4 phosphorothioates at the 3 end 3 end cholesterol group and full-length nucleotide 2 -methoxy modification They are designed to mimic endogenous miRNAs and recommended for miRNA functional studies Compared with miRNA mimics they exhibits enhanced cellular uptake stability and regulatory activity in vivo
Catalog No. 50-003-22450