Learn More
Sigma Aldrich Fine Chemicals Biosciences HSf1 Activator HSf1A - CAS
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 5337610001
Encompass
HSf1 Activator HSf1A CAS 1196723-93-9 is a cell-permeable activator of HSf1 that protects mammalian cells against stress-induced apoptosis. Protects cells from polyQ cytotoxicity.
Catalog No. 50-289-9056