Learn More
Cayman Chemical Hsf1humnrecombInt 100ug
SDP
Supplier:  Cayman Chemical 40253100 UG
Encompass_Preferred
Source: Recombinant human N-Terminal His-tagged Hsf1 expressed in E. coli • Amino acids: Met1; 2-529 (full length) • MW: 58.6 kDa
Catalog No. 50-294-4821