Learn More
TARGETMOL CHEMICALS INC hSMG-1 inhibitor 11j 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T88841MG
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. hSMG-1 inhibitor 11j is a pyrimidine derivative. hSMG-1 inhibitor 11j is a potent and selective inhibitor of hSMG-1, IC50 = 0.11 nM. hSMG-1 inhibitor 11j exhibits >455-fold selectivity for hSMG-1 over mTOR (IC50=50 nM), PI3Kalpha/gamma (IC50=92/60 nM) and CDK1/CDK2 (IC50=32/7.1 uM). hSMG-1 inhibitor 11j can be used for the research of cancer. Purity 99.29%
Catalog No. 50-288-0793