Learn More
TARGETMOL CHEMICALS INC HSP27 inhibitor J2 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T72651MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. HSP27 inhibitor J2 (J2) (J2) is a HSP27 inhibitor, inhibits a production of HSP27 giant polymers, thereby having an effect of inhibiting a chaperone function of the HSP27 and reducing a cell protection function. Purity 98%
Catalog No. 50-287-9250