Learn More
Sino Biological Peptide Substrates: HSP27tide
Supplier:  Sino Biological H3158
Encompass_Preferred
The HSP27tide peptide sequence (RRLNRQLSVA-amide) is based on the mouse HSP27 (amino acid 80-85).
Catalog No. 50-256-9033