Learn More
ABclonal Technology HSP40 Rabbit mAb
SDP
Supplier:  ABclonal Technology A23957100UL
Encompass_Preferred
This gene encodes a member of the DnaJ or Hsp40 (heat shock protein 40 kD) family of proteins DNAJ family members are characterized by a highly conserved amino acid stretch called the J-domain and function as one of the two major classes of molecular chaperones involved in a wide range of cellular events such as protein folding and oligomeric protein complex assembly The encoded protein is a molecular chaperone that stimulates the ATPase activity of Hsp70 heat-shock proteins in order to promote protein folding and prevent misfolded protein aggregation Alternative splicing results in multiple transcript variants
Catalog No. 50-000-06063