Learn More
Enzo Life Sciences HSP 70 HSP 72 polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA812D
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human Hsp70. UniProt: P0DMV8 (HSPA1A), P0DMV9 (HSPA1B). Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Sheep, Porcine, Monkey, Hamster, Guinea pig, Fish, Drosophila, Dog, Bovine, Beluga. Applications: ICC, IF, IHC, IP, WB. Recommended dilutions: Immunoprecipitation (1:100). Western Blot (1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-563