Learn More
Medchemexpress LLC Hsp90-IN-17 hydrochloride | 1253584-63-2 | 99.3% | 476.87 | 100 MG

Supplier: Medchemexpress LLC HY148215A100MG
Hsp90-IN-17 hydrochloride is an HSP90 inhibitor that can be used in the study of proliferative diseases, such as cancer and neurodegenerative diseases.
- Potent HSP90 inhibitor
- Useful for studying proliferative diseases
- Shows antiproliferative activity against human A2780 cells
- Provided as a solid, light yellow to yellow in color
- Recommended storage at 4°C, sealed, away from moisture and light
- Stable in solvent at -80°C for 6 months, or -20°C for 1 month
- Soluble in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.