Learn More
Medchemexpress LLC Hsp90-IN-17 hydrochloride | 1253584-63-2 | 99.3% | 476.87 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY148215A5MG
Encompass_Preferred
Hsp90-IN-17 (Example 5) hydrochloride is an HSP90 inhibitor that can be used in the study of proliferative diseases, such as cancer and neurodegenerative diseases. It has demonstrated antiproliferative activity against human A2780 cells with an IC50 of 0.362 μM after 72 hours.
  • HSP90 inhibitor.
  • Used in the study of proliferative diseases.
  • Applicable to research on cancer and neurodegenerative diseases.
  • Exhibits antiproliferative activity.
Catalog No. 50-003-61451