Learn More
Medchemexpress LLC Hsp90-in-17 hydrochloride | 1253584-63-2 | 99.31% | 476.87 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY148215A
Encompass_Preferred
Hsp90-IN-17 hydrochloride is an HSP90 inhibitor that can be used in the study of proliferative diseases, such as cancer and neurodegenerative diseases.
  • HSP90 inhibitor
  • Used in the study of proliferative diseases
  • Suitable for cancer research
  • Suitable for neurodegenerative diseases research
Catalog No. 50-003-63474