Learn More
Medchemexpress LLC HSP90AA1 Human His 2ug
SDP
Supplier:  Medchemexpress LLC HYP722382UG
Encompass_Preferred
The HSP90AA1 protein is an important molecular chaperone that coordinates the maturation and regulation of specific target proteins critical for cell cycle control and signal transduction Its ATPase activity drives a functional cycle that induces conformational changes in client proteins for activation HSP90AA1 Protein Human (His-SUMO) is the recombinant human-derived HSP90AA1 protein expressed by E coli with N-6 His N-SUMO labeled tag
Catalog No. 50-001-82584