Learn More
Aviva Systems Biology Corp HSPA1A Antibody - middle region (ARP33096_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33096_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPA1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSPA1A. Alternate names: HSP72, HSPA1, HSP70I, HSPA1B, HSP70-1, HSP70-1AApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54881