Learn More
Waters Corp HSPgel Column, 6 um, 6 mm x 150 mm
Supplier: Waters Corp 186001787
The HSPgel Column, AQ 4 is a Size Exclusion Chromatography (SEC) / Aqueous Size Exclusion Chromatography (GFC) column. It is designed for the separation of macromolecules in aqueous mobile phases.
- Particle size: 6 μm
- Column dimensions: 6.0 mm internal diameter x 150 mm length
- Molecular weight range: 10,000 to 400,000 (PEG/PEO)
- Chemistry producers term: AQ 4.0
- USP Class: L38
- Technology: HPLC
- Matrix: Polymer
- Particle morphology: Spherical, fully porous
- Recommended operating temperature: Ambient to 80 °C
- Recommended maximum injection volume: 20 μL
- Typical injection volume: 1 μL
- Theoretical plates: 10,000
- Parker style endfittings with 0.090 in. ferrule depth
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.