Learn More
Waters Corp Waters HSPgel Column, 3 μm, 6 mm X 150 mm, 1K to 20K
Supplier: Waters Corp 186001743
HSPgel columns are designed for high-speed GPC analysis, providing fast, accurate, and precise molecular weight polymer determinations. They increase sample throughput and reduce solvent consumption, with analysis times under 7 minutes and solvent usage below 5 mL per run, optimizing performance with lower volume columns.
- Separation mode: SEC/GPC
- Particle substrate: Polymer
- Molecular weight range: 1,000 to 20,000
- Particle shape: Spherical
- Particle size: 3 μm
- Endfitting type: Parker-style
- Format: Column
- Packing solvent: THF
- System: HPLC
- Technique: GPC
- Inner diameter: 6 mm
- Length: 150 mm
- Units per package: 1 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.