Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp HSPgel HT Column, 5 μm, 50K–10M MW, ODCB

Supplier:  Waters Corp 186001779

Encompass_Preferred

HSPgel HT series columns are designed for Gel Permeation Chromatography (GPC) analysis of organic polymers. These columns can operate across a wide temperature range, from room temperature up to 180°C. They are shipped in o-dichlorobenzene (ODCB) and allow for multiple solvent transfers.

  • Particle size: 5 μm
  • Molecular weight range: 50,000–10,000,000 (based on polystyrene chain lengths)
  • Column dimensions: 6.0 x 150 mm
  • Recommended flow rate range: 0.30–0.70 mL/min
  • Maximum flow rate: 0.80 mL/min
  • Maximum pressure drop: 415 PSI
  • Operating temperature range: Ambient to 180°C
  • Maximum injection volume: 20 μL
  • Theoretical plates: 8,000
  • Features Parker style endfittings with a 0.090 in. ferrule depth

Catalog No. 50-780-750


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.