Learn More
Waters Corp HSPgel HT MB-L Column, 5 µm, 6 mm X 150 mm, 100 - 1K, THF, 1/pk
Supplier:  Waters Corp 186001769
Encompass_Preferred
The HSPgel columns are specifically designed for high speed GPC analysis. They offer fast, accurate and precise molecular weight polymer determinations as well as increased sample throughput and greatly reduced solvent consumption and disposal costs. With the advent of today's higher performance solvent management systems, many polymers can now be analyzed accurately and reproducibly utilizing lower volume columns. This results in total analysis times of 7 minutes or less, thereby increasing sample throughput up to 6x while decreasing solvent usage from 35-50 mL to 5 mL or less per run.
Catalog No. 50-780-740