Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Column, 3 μm Particle, 6 mm x 150 mm, 2K–60K MW

Supplier:  Waters Corp 186001752

Encompass_Preferred

HSPgel columns are designed for high-speed GPC analysis, offering fast, accurate, and precise molecular weight polymer determinations. They increase sample throughput and reduce solvent consumption, allowing total analysis times of 7 minutes or less and decreasing solvent usage from 35–50 mL to 5 mL or less per run.

  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • Molecular weight range: 2000 to 60000
  • Particle shape: Spherical
  • Particle size: 3 μm
  • Endfitting type: Parker-style
  • Packing solvent: THF
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 6 mm
  • Length: 150 mm

Catalog No. 50-780-724


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.