Learn More
Waters Corp Waters XSelect HSS C18 Column, 3.5 μm, 100 Å, 3 mm X 150 mm
Supplier:  Waters Corp 186004763
Encompass_Preferred
This general-purpose C18 bonded phase column utilizes HSS (High Strength Silica) technology, providing superior peak shape for bases and increased retention. It is designed for exceptional lifetimes at low pH and can sustain UPLC pressures.
  • Provides superior peak shape for bases.
  • Offers increased retention.
  • Exceptional column lifetimes at low pH.
  • Selectivity and retention similar to modern, fully endcapped, high-coverage silica-based C18 columns.
  • Utilizes HSS (High Strength Silica) technology.
  • Complements chromatographic performance of BEH and CSH particles.
  • Sustains UPLC pressures.
  • Fully scalable and transferable to UPLC separations.
  • Delivers benefits including increased polar compound retention and different selectivity.
  • Recommended operational pH range: 1 to 8.
  • Designed to withstand a pressure of 6000 psi (415 Bar).
Catalog No. 50-466-080