Learn More
Waters Corp XSelect Column, HSS C18, 100Å, 3.5 μm, 3 mm × 100 mm
Supplier:  Waters Corp 186004762
Encompass_Preferred
The XSelect HSS C18 Column is a general-purpose C18 bonded phase column that offers excellent peak shape for bases, increased retention, and exceptional column lifetimes at low pH. This high-performance, reversed-phase column is designed to separate a wide range of compounds, making it suitable for applications in pharmaceuticals, biomolecules, food and beverage, environmental analysis, forensic analysis, quality control, and research.
  • Superior peak shape for bases.
  • Increased retention.
  • Exceptional column lifetimes at low pH.
  • Exceptional separation efficiency and fast separations.
  • Enhanced selectivity for basic compounds through proprietary HSS C18 chemistry.
  • Compatibility with a diverse range of HPLC systems.
  • High-purity construction ensures no contaminants interfere with separations.
  • Pre-conditioned, enabling immediate usage upon receipt.
Catalog No. 50-466-079