Learn More
Waters Corp Waters XSelect Column, HSS C18, 100Å, 5 μm, 4.6 mm, 250 mm
Supplier:  Waters Corp 186004775
Encompass_Preferred
The HSS C18 is a general-purpose C18 bonded phase column designed to provide superior peak shape for bases, increased retention compared to XBridge C18 columns, and exceptional column lifetimes at low pH. The selectivity and retention characteristics of these columns are comparable to most modern, fully endcapped, high-coverage silica-based C18 columns.
  • Enhances chromatographic performance through High Strength Silica (HSS) technology.
  • Withstands UPLC pressures, making it scalable and transferable to UPLC separations.
  • Provides improved polar compound separation and different selectivity.
  • Isolates and quantifies sample constituents with high resolution for complex samples.
  • Operates within a pH range of 1 to 8 and withstands pressures up to 6000 psi (415 bar).
Catalog No. 50-466-092