Learn More
Waters Corp XSelect HSS C18 SB Column, 100Å, 3.5 µm, 4.6 mm X 250 mm, 1/pk
Supplier:  Waters Corp 186004751
Encompass_Preferred
The HSS C18 SB (Selectivity for Bases) is an unendcapped, low-coverage C18 bonded-phase column that was designed specifically to provide increased silanophilic interactions. This results in an alternate selectivity compared to a high-coverage fully-endcapped C18 column, particularly for basic compounds.
Catalog No. 50-466-068