Learn More
Waters Corp Waters Column, C18 SB, 5 μm, 4.6 mm x 250 mm
Supplier:  Waters Corp 186004756
Encompass_Preferred
The HSS C18 SB column is unendcapped, low-coverage C18 bonded-phase, providing increased silanophilic interactions. It offers alternate selectivity for basic compounds, separating complex mixtures at low pH for improved resolution. HSS technology enhances polar molecule retention and handles UPLC pressures.
  • Low coverage, non-end-capped C18 phase.
  • Increased silanophilic interactions for alternate selectivity.
  • Enhanced peak shape.
  • Minimal column bleed and excellent consistency.
  • Ultra-efficiencies and simple format conversion.
  • Improved resolution for complex samples.
  • Increased selectivity and retention with low-pH mobile phases.
  • HSS technology for enhanced polar molecule retention, selectivity, and UPLC pressure tolerance.
  • Fully scalable for UPLC.
Catalog No. 50-466-073