Learn More
TYRELL MEDICAL LLC Dielectric High-Performance Heat Transfer Fluid 5kg 4/CS SEA FRT
Supplier:  TYRELL MEDICAL LLC TCHT135 5KG 4/CS SEA
Encompass_Preferred
Dielectric High-Performance Heat Transfer Fluid 5kg 4/CS SEA FRT. Heat conduction of machinery and equipment, electronics cooling and cooling of datacenter equipment. CAS Number 69991-67-9. Melting Point / Freezing Point <-90 °C (<-130 °F), Initial boiling point and boiling range 125-132 C (257-296.6 F).
Catalog No. 502120242