Learn More
Wilmad Labglass HT MNTL HEMI 125ML
Supplier:  Wilmad Labglass LG8800104
Encompass
Heating Mantle, Hemispherical. Size: 125mL. Watts: 80. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.
Catalog No. 50-801-161
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.