Learn More
Wilmad Labglass HT MNTL KTTL AL HOUSING 2000ML
Supplier:  Wilmad Labglass LG8883106
Encompass
Heating Mantle, Reaction Kettle, Aluminum Housing. Size: 2000mL; Watts: 470. Volts: 115. Aluminum housing exterior-designed to be used w/ reaction vessels LG-8071- through LG-8082. Supplied w/ 3-Wire cord and locking connector.
Catalog No. 50-801-097
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.