Learn More
HTDialysis LLC 12-14kDa MWCO regenerated cellulose dialysis membrane strips. Package contains 50 individual dialysis strips for up to 600 assays. Designed for HTD reusable micro equilibrium dialysis or buffer exchange and desalting devices.
Supplier:  HTDialysis LLC 1101
Encompass
Custom cut regenerated cellulose dialysis membrane strips. Designed for HTD reusable micro equilibrium dialysis or desalting devices Features- Range of pore sizes: 1kDa, 3.5kDa, 6-8kDa, 10kDa, 12-14kDa, 25kDa, 50kDa Applications- Serum/plasma protein binding- Microsome protein binding- Relative binding of drug to specific tissues- Receptor binding- T3/T4 free fraction binding (hyperthyroidism) determinations- All desalting and buffer exchange steps of sample processing applications: dialysis of proteins, oligonucleotides, RNA or DNA samples- Removal of sample contaminates - salts, solvents, detergents- Conducting Formation studies - protein-protein, protein-DNA, protein-RNAFor use with:HTD96B – equilibrium dialysis device HTD96C – equilibrium dialysis deviceHTD12A – Buffer exchange and desalting device
Catalog No. NC9076208