Learn More
Selleck Chemical LLC HTH-01-015
SDP
Supplier:  Selleck Chemical LLC S731810MM/1ML
Encompass_Preferred
HTH-01-015 is a potent and selective NUAK1 inhibitor with IC50 of 100 nM 100-fold selectivity over NUAK2
Catalog No. 50-000-04671