Learn More
Medchemexpress LLC HY-100768 10mg Medchemexpress, HTHQ CAS:148081-72-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10076810MG
Encompass_Preferred
Medchemexpress, HY-100768 10mg HTHQ CAS:148081-72-5 HTHQ (1-O-hexyl-2,3,5-trimethylhydroquinone) is a potent lipophilic phenolic antioxidant. HTHQ has considerable anti-oxidative activity by directly reacting with reactive oxygen species (ROS) and scavenging ROS to form more stable free radicals [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1942