Learn More
Apexbio Technology LLC HTHQ 148081-72-5 50mg
SDP
Supplier:  Apexbio Technology LLC BA621150
Encompass_Preferred
HTHQ (1-O-hexyl-2 3 5-trimethylhydroquinone CAS 148081-72-5) is a lipophilic phenolic compound that exhibits potent antioxidant activity It exerts its effect by directly reacting with reactive oxygen species (ROS) thereby neutralizing them and forming more stable radical species Through this mechanism HTHQ mitigates oxidative stress in biological systems This compound is utilized in biomedical research to investigate oxidative damage redox biology and cellular protection mechanisms associated with lipid peroxidation and free radical-mediated cellular injury
Catalog No. 50-000-87934