Learn More
Bon Opus Biosciences Recombinant Human/Mouse Activin A/INHBA 1mg
SDP
Supplier:  Bon Opus Biosciences C6871MG
Encompass_Preferred
Recombinant Human Activin A is produced by our Mammalian expression system and the target gene encoding Gly311-Ser426 is expressed. Accession# P08476. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# C687
Catalog No. 50-217-2756