Learn More
Bon Opus Biosciences Recombinant Human Angiopoietin-related protein 4/ANGPTL4 (N-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP107500UG
Encompass_Preferred
Recombinant Human Angiopoietin-related protein 4 is produced by Human 293 Cells. The target gene encoding P166-S406 is expressed with a 6His tag at the N terminus. Expression system: Human 293 Cells. Accession# Q9BY76. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP107
Catalog No. 50-208-0182